Learn More
Socorex™ Laboratory Pipette Racks
Hold and organize pipettes using Socorex™ Laboratory Pipette Stands. The space-saving work stations and shelf holder for pipettes come in a variety of colors. All are constructed of sturdy polyamid or polycarbonate materials and are very stable with anti-slip pads.
195.00 DKK - 933.00 DKK
Product Code | Brand | dimColor | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | dimColor | Price | Quantity & Availability | ||||||
10201852
|
Socorex™
320.340 |
933.00 DKK
Each |
Estimated Shipment: 15-09-2025
Log in to see stock. |
|||||||
10274591
|
Socorex™
320.337B |
933.00 DKK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||||
10266982
|
Socorex™
320.337R |
933.00 DKK
Each |
Estimated Shipment: 22-09-2025
Log in to see stock. |
|||||||
10527854
|
Socorex™
320.332 |
195.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||||
Description
- Especially suited for single- and multi-channel pipettes and macropipettes.
- Accommodate all Calibra™ and Acura™ models
- Available in pastel colors
- Constructed of easy-to-clean polyamid and polycarbonate material