Learn More
Socorex™ Laboratory Pipette Racks
Hold and organize pipettes using Socorex™ Laboratory Pipette Stands. The space-saving work stations and shelf holder for pipettes come in a variety of colors. All are constructed of sturdy polyamid or polycarbonate materials and are very stable with anti-slip pads.
Brand: Socorex™ 320.337R
This item is not returnable.
View return policy
Description
- Especially suited for single- and multi-channel pipettes and macropipettes.
- Accommodate all Calibra™ and Acura™ models
- Available in pastel colors
- Constructed of easy-to-clean polyamid and polycarbonate material
Specifications
Universal Workstation | |
Model 337 Universal Work Station | |
Single-channel Micropipettes | |
7 |
Product Content Correction
Product Title
Socorex™ Laboratory Pipette Racks > Color: Pastel rose
Spot an opportunity for improvement?Share a Content Correction