Learn More
Cole-Parmer™ PFA Female Threaded Tee Fitting
Threaded fittings offer more permanent installation
730.00 DKK - 1670.00 DKK
Specifications
Product Type | Female Threaded Tee |
---|---|
Description | NPT(F) Connection |
Autoclavable | Not Autoclavable |
Material | PFA |
Sterilization Method | Ethylene Oxide |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
11838043
|
Cole-Parmer™
06546-20 |
1/2 in. |
1135.00 DKK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11828043
|
Cole-Parmer™
06546-19 |
3/8 in. |
730.00 DKK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
11848043
|
Cole-Parmer™
06546-21 |
3/4 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Chemically-inert PFA handles most aggressive chemicals
- Threaded reducing bushing
- Choose the size and material that meets your application needs
- Use with our rigid tubing for your high-pressure applications
Specifications
Female Threaded Tee | |
Not Autoclavable | |
Ethylene Oxide | |
Translucent |
NPT(F) Connection | |
PFA | |
T-shape |