Learn More
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30018661
|
Melford
P20250-500.0 |
500 g |
872.00 DKK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018662
|
Melford
P20250-5000.0 |
5 kg |
7025.00 DKK
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018660
|
Melford
P20250-1000.0 |
1 kg |
1495.00 DKK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mixtures of peptides and free amino acids from peptic digest of animal tissue.
Mixtures of peptides and free amino acids from peptic digest of animal tissue.- Amino Nitrogen (AN): 2.5-3.5%
- Loss on Drying: <6.0%
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Melford Peptone, Granulated