Learn More
Melford Peptone, Granulated
Mixtures of peptides and free amino acids from peptic digest of animal tissue.
872.00 DKK - 7025.00 DKK
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30018661
View Documents
Melford
P20250-500.0
500 g
872.00 DKK
500g
View alternative products
Save Up to 
»
30018662
View Documents
Melford
P20250-5000.0
5 kg
7025.00 DKK
5kg
View alternative products
Save Up to 
»
30018660
View Documents
Melford
P20250-1000.0
1 kg
1495.00 DKK
1kg
View alternative products
Save Up to 
»
Description
Description
Mixtures of peptides and free amino acids from peptic digest of animal tissue.
Mixtures of peptides and free amino acids from peptic digest of animal tissue.
  • Amino Nitrogen (AN): 2.5-3.5%
  • Loss on Drying: <6.0%
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Melford Peptone, Granulated