Learn More
3M™ PELTOR™ Bull's Eye™ I Earmuffs
3M™ PELTOR™ Bull's Eye™ I Earmuffs ensure protection in high noise environments without being too bulky.
3451.16 DKK - 5335.00 DKK
Specifications
Certifications/Compliance | CE Approved , EN 352-1: 2002 |
---|---|
Noise Reduction Rating (NRR) | 27 dB |
Product Type | Ear Muffs |
Style | Foldable, over the head |
Foldable | Yes |
Description
- Shaped to avoid interference with the rifle butt and not distract when shooting
- Soft wide cushions helps reduce pressure around the ears and improves comfort and wearability
- Easy to replace cushions and inserts helps keep them hygienically clean
- Collapsible headband for easy and protective storage in a pocket or bag
- Attenuation Rating:
- High: 32
- Medium: 24
- Low: 15
Specifications
CE Approved , EN 352-1: 2002 | |
Ear Muffs | |
Yes |
27 dB | |
Foldable, over the head |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
3M™ PELTOR™ Bull's Eye™ I Earmuffs