Learn More
Orantinib, MedChemExpress
MedChemExpress Orantinib (SU6668; TSU-68) is a multi-targeted receptor tyrosine kinase inhibitor with Kis of 2.1 μM, 8 nM and 1.2 μM for Flt-1, PDGFRβ and FGFR1, respectively.

936.63 DKK - 6757.79 DKK
Specifications
Chemical Name or Material | Orantinib |
---|---|
Physical Form | Powder |
CAS | 252916-29-3 |
Color | Deep Yellow |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16474018
![]() |
Medchem Express
HY-10517 |
10 mg |
936.63 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16420377
![]() |
Medchem Express
HY-10517 |
50 mg |
3958.00 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16415016
![]() |
Medchem Express
HY-10517 |
100 mg |
6757.79 DKK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Orantinib (SU6668Specifications
Orantinib | |
252916-29-3 | |
Research | |
Cancer-Kinase/protease | |
DMSO : ≥ 28 mg/mL (90.22 mM) | |
310.35 | |
98.18% |
Powder | |
Deep Yellow | |
C18H18N2O3 | |
SU6668 TSU-68 | |
CC1=C(/C=C2C3=CC=CC=C3NC/2=O)NC(C)=C1CCC(O)=O | |
310.35 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.