missing translation for 'onlineSavingsMsg'
Learn More
Orantinib, MedChemExpress Product Code.: 16420377

Orantinib, MedChemExpress

Product Code. 16420377
50 mg, 50mg
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16420377

Brand: Medchem Express HY10517

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Orantinib (SU6668; TSU-68) is a multi-targeted receptor tyrosine kinase inhibitor with Kis of 2.1 μM, 8 nM and 1.2 μM for Flt-1, PDGFRβ and FGFR1, respectively.

Orantinib (SU6668

Specifications

Chemical Name or Material Orantinib
Physical Form Powder
CAS 252916-29-3
Color Deep Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H18N2O3
For Use With (Application) Cancer-Kinase/protease
Synonym SU6668 TSU-68
Solubility Information DMSO : ≥ 28 mg/mL (90.22 mM)
SMILES CC1=C(/C=C2C3=CC=CC=C3NC/2=O)NC(C)=C1CCC(O)=O
Molecular Weight (g/mol) 310.35
Formula Weight 310.35
Percent Purity 98.18%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Orantinib, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.