Learn More
Ansell MICROCHEM™ 6000
Protective suit with multi-layer chemical barrier technology for Type 1 / Level A Gas-tight protection.
10315.00 DKK - 19355.00 DKK
Specifications
Color | Orange |
---|---|
For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases, Tank Cleaning |
Weight (Metric) | 230 gsm |
Product Code | Brand | Size | Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Style | Price | Quantity & Availability | |||||
17100915
|
Ansell
OR60T-00801-04-G02 |
Large | O60T801B | N/A | ||||||
17100925
|
Ansell
OR60T-00803-02-GA1 |
Small | O60T803E | N/A | ||||||
17100945
|
Ansell
OR60T-00809-04-GA1 |
Large | O60T809E | N/A | ||||||
17100955
|
Ansell
OR60TA00812-02-GA1 |
Small | O60A812E | N/A | ||||||
17100965
|
Ansell
OR60TA00813-06-GA1 |
2X-Large | O60A813E | N/A | ||||||
17100975
|
Ansell
OR60TA00818-04-GA1 |
Large | O60A818E | N/A | ||||||
17110915
|
Ansell
OR60T-00801-05-G02 |
X-Large | O60T801B | N/A | ||||||
17110925
|
Ansell
OR60T-00803-03-GA1 |
Medium | O60T803E | N/A | ||||||
17110945
|
Ansell
OR60T-00809-05-GA1 |
X-Large | O60T809E | N/A | ||||||
17110955
|
Ansell
OR60TA00812-03-GA1 |
Medium | O60A812E | N/A | ||||||
17110965
|
Ansell
OR60TA00813-07-GA1 |
3X-Large | O60A813E | N/A | ||||||
17110975
|
Ansell
OR60TA00818-05-GA1 |
X-Large | O60A818E | N/A | ||||||
17120915
|
Ansell
OR60T-00801-06-G02 |
2X-Large | O60T801B | N/A | ||||||
17120925
|
Ansell
OR60T-00803-04-GA1 |
Large | O60T803E | N/A | ||||||
17120935
|
Ansell
OR60T-00808-02-GA1 |
Small | O60T808E | N/A | ||||||
17120945
|
Ansell
OR60T-00809-06-GA1 |
2X-Large | O60T809E | N/A | ||||||
17120955
|
Ansell
OR60TA00812-04-GA1 |
Large | O60A812E | N/A | ||||||
17120975
|
Ansell
OR60TA00818-06-GA1 |
2X-Large | O60A818E | N/A | ||||||
17130915
|
Ansell
OR60T-00801-07-G02 |
3X-Large | O60T801B | N/A | ||||||
17130925
|
Ansell
OR60T-00803-05-GA1 |
X-Large | O60T803E | N/A | ||||||
17130935
|
Ansell
OR60T-00808-03-GA1 |
Medium | O60T808E | N/A | ||||||
17130945
|
Ansell
OR60T-00809-07-GA1 |
3X-Large | O60T809E | N/A | ||||||
17130955
|
Ansell
OR60TA00812-05-GA1 |
X-Large | O60A812E | N/A | ||||||
17130975
|
Ansell
OR60TA00818-07-GA1 |
3X-Large | O60A818E | N/A | ||||||
17140915
|
Ansell
OR60T-00802-02-GA1 |
Small | O60T802E | N/A | ||||||
17140925
|
Ansell
OR60T-00803-06-GA1 |
2X-Large | O60T803E | N/A | ||||||
17140935
|
Ansell
OR60T-00808-04-GA1 |
Large | O60T808E | N/A | ||||||
17140955
|
Ansell
OR60TA00812-06-GA1 |
2X-Large | O60A812E | N/A | ||||||
17140975
|
Ansell
OR60TA00819-02-GA1 |
Small | O60A819E | N/A | ||||||
17150915
|
Ansell
OR60T-00802-03-GA1 |
Medium | O60T802E | N/A | ||||||
17150925
|
Ansell
OR60T-00803-07-GA1 |
3X-Large | O60T803E | N/A | ||||||
17150935
|
Ansell
OR60T-00808-05-GA1 |
X-Large | O60T808E | N/A | ||||||
17150955
|
Ansell
OR60TA00812-07-GA1 |
3X-Large | O60A812E | N/A | ||||||
17150975
|
Ansell
OR60TA00819-03-GA1 |
Medium | O60A819E | N/A | ||||||
17160915
|
Ansell
OR60T-00802-04-GA1 |
Large | O60T802E | N/A | ||||||
17160935
|
Ansell
OR60T-00808-06-GA1 |
2X-Large | O60T808E | N/A | ||||||
17160955
|
Ansell
OR60TA00813-02-GA1 |
Small | O60A813E | N/A | ||||||
17160975
|
Ansell
OR60TA00819-04-GA1 |
Large | O60A819E | N/A | ||||||
17170915
|
Ansell
OR60T-00802-05-GA1 |
X-Large | O60T802E | N/A | ||||||
17170935
|
Ansell
OR60T-00808-07-GA1 |
3X-Large | O60T808E | N/A | ||||||
17170955
|
Ansell
OR60TA00813-03-GA1 |
Medium | O60A813E | N/A | ||||||
17170975
|
Ansell
OR60TA00819-05-GA1 |
X-Large | O60A819E | N/A | ||||||
17180905
|
Ansell
OR60T-00801-02-G02 |
Small | O60T801B | N/A | ||||||
17180915
|
Ansell
OR60T-00802-06-GA1 |
2X-Large | O60T802E | N/A | ||||||
17180935
|
Ansell
OR60T-00809-02-GA1 |
Small | O60T809E | N/A | ||||||
17180955
|
Ansell
OR60TA00813-04-GA1 |
Large | O60A813E | N/A | ||||||
17180965
|
Ansell
OR60TA00818-02-GA1 |
Small | O60A818E | N/A | ||||||
17180975
|
Ansell
OR60TA00819-06-GA1 |
2X-Large | O60A819E | N/A | ||||||
17190905
|
Ansell
OR60T-00801-03-G02 |
Medium | O60T801B | N/A | ||||||
17190915
|
Ansell
OR60T-00802-07-GA1 |
3X-Large | O60T802E | N/A | ||||||
17190935
|
Ansell
OR60T-00809-03-GA1 |
Medium | O60T809E | N/A | ||||||
17190955
|
Ansell
OR60TA00813-05-GA1 |
X-Large | O60A813E | N/A | ||||||
17190965
|
Ansell
OR60TF00818-03-GA1 |
Medium | O60A818E | N/A | ||||||
17190975
|
Ansell
OR60TA00819-07-GA1 |
3X-Large | O60A819E | N/A | ||||||
Description
- Excellent chemical protection, >480 mins for 15 chemicals specified by EN 943-2:2002
- Innovative fabric technology, high performance barrier laminate with scrim reinforcement
- Lightweight and flexible
- Highly visible bright orange color for improved worker safety
- Ultrasonically welded and hot air taped seams
- DYNAT/YKK Gas-Tight zipper provides protection and performance in the most hostile of chemical environments
- The zip has an outer zip flap made of the same material as the suit
- Semi-rigid 3 layer GAG visor provides excellent optical clarity
Specifications
Orange | |
Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases, Tank Cleaning | |
230 gsm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell MICROCHEM™ 6000