Få mere at vide
Ansell MICROCHEM™ 6000
Protective suit with multi-layer chemical barrier technology for Type 1 / Level A Gas-tight protection.
Brand: Ansell OR60T-00808-02-GA1
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
- Excellent chemical protection, >480 mins for 15 chemicals specified by EN 943-2:2002
- Innovative fabric technology, high performance barrier laminate with scrim reinforcement
- Lightweight and flexible
- Highly visible bright orange color for improved worker safety
- Ultrasonically welded and hot air taped seams
- DYNAT/YKK Gas-Tight zipper provides protection and performance in the most hostile of chemical environments
- The zip has an outer zip flap made of the same material as the suit
- Semi-rigid 3 layer GAG visor provides excellent optical clarity
Tekniske data
Orange | |
Multi-Layer Non-Woven Barrier Laminate Fabric | |
Welded and Taped | |
230 gsm |
Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases, Tank Cleaning | |
Small | |
O60T808E |
Korrektion af produktindhold
Produkttitel
Ansell MICROCHEM™ 6000 > Size: Small, Style: O60T808E
Ser du en mulighed for forbedring?Del en indholdskorrektion