Learn More
Ansell MICROCHEM™ 6000
Protective suit with multi-layer chemical barrier technology for Type 1 / Level A Gas-tight protection.
Brand:  Ansell OR60TA00812-04-GA1
 View more versions of this product
Product Code. 17120955
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Excellent chemical protection, >480 mins for 15 chemicals specified by EN 943-2:2002
  • Innovative fabric technology, high performance barrier laminate with scrim reinforcement
  • Lightweight and flexible
  • Highly visible bright orange color for improved worker safety
  • Ultrasonically welded and hot air taped seams
  • DYNAT/YKK Gas-Tight zipper provides protection and performance in the most hostile of chemical environments
  • The zip has an outer zip flap made of the same material as the suit
  • Semi-rigid 3 layer GAG visor provides excellent optical clarity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Orange
Multi-Layer Non-Woven Barrier Laminate Fabric
Welded and Taped
230 gsm
Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases, Tank Cleaning
Large
O60A812E
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Ansell MICROCHEM™ 6000 > Size: Large, Style: O60A812E