Learn More
Hirschmann™ Measuring flask Class A with pre-packing regulation
Achieve precise standard solutions. Hirschmann™ Measuring flask Class A with pre-packing regulation is standardized to contain or to deliver a definite amount of liquid. Flask has blue graduations.
1015.00 DKK - 1690.00 DKK
Product Code | Brand | Graduation Range | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Graduation Range | Price | Quantity & Availability | ||||||
11893212
|
Hirschmann™
2950188 |
323-337 mL |
1540.00 DKK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
11823212
|
Hirschmann™
2950165 |
18 to 22 mL |
1175.00 DKK
Pack of 2 |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
11692229
|
Hirschmann™
2950184 |
368-382 mL |
1550.00 DKK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
11833212
|
Hirschmann™
2950170 |
23 to 27 mL |
1175.00 DKK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
11892582
|
Hirschmann™
2950175 |
47 to 53 mL |
1260.00 DKK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
11873212
|
Hirschmann™
2950186 |
245 to 255 mL |
1435.00 DKK
Pack of 2 |
|
||||||
11803222
|
Hirschmann™
2950189 |
343 to 357 mL |
1540.00 DKK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
11813222
|
Hirschmann™
2950190 |
490 to 510 mL |
1690.00 DKK
Pack of 2 |
In Stock
Log in to see stock. ![]() |
||||||
10421961
|
Hirschmann™
2950191 |
690 to 710 mL |
1015.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- Class A
- Pursuant to the rules of the pre-packing regulation
- Certified conformity
- Blue graduation
- Non-ground, rimmed
- Printed scale on the neck for simple and rapid identification of deviation of actual value from normal value
- Calibrated for IN
Specifications
Yes | |
A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hirschmann™ Measuring flask Class A with pre-packing regulation