Learn More
DWK Life Sciences Kimble™ KIMAX™ Erlenmeyer Flasks with Screw Caps
Plastic-coated for added safety
2915.00 DKK - 5687.51 DKK
Specifications
Closure Color | Blue |
---|---|
Closure Material | Polypropylene |
Description | Reverse metric scale graduations |
Material | Borosilicate Glass |
Includes | Clear pour rings |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11787979
|
DWK Life Sciences
KC26720-250 |
250 mL |
4480.00 DKK
Pack of 6 |
Estimated Shipment: 10-10-2025
Log in to see stock. |
||||||
11797979
|
DWK Life Sciences
KC26720-500 |
500 mL |
5200.00 DKK
Pack of 6 |
Estimated Shipment: 03-09-2025
Log in to see stock. |
||||||
11707989
|
DWK Life Sciences
KC267201000 |
1000 mL | N/A | |||||||
11717989
|
DWK Life Sciences
KC26720-2000 |
2000 mL |
2915.00 DKK
Pack of 2 |
Estimated Shipment: 03-09-2025
Log in to see stock. |
||||||
Description
Plastic-coated for added safety
- KimCote™ plastic coating
- Large openings and GL 45 threaded closures for easy access and tight sealing
- Blue caps and clear pour rings included
- Marking spots and scales are durable white ceramic enamel
Specifications
Blue | |
Reverse metric scale graduations | |
Clear pour rings | |
Screw |
Polypropylene | |
Borosilicate Glass | |
White Ceramic Enamel Marking |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
DWK Life Sciences Kimble™ KIMAX™ Erlenmeyer Flasks with Screw Caps