Learn More
DWK Life Sciences Kimble™ Headspace Vials
Glass, Clear KG-33
5795.00 DKK - 7020.00 DKK
Specifications
Color | Clear |
---|---|
Diameter (Metric) Outer | 23 mm |
Material | Silica Glass |
No. per Case | 1000 |
Quantity | 1000 Cs. |
Product Code | Brand | Closure Type | Height (Metric) | Volume (Metric) | Description | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Type | Height (Metric) | Volume (Metric) | Description | Price | Quantity & Availability | ||
11883023
|
DWK Life Sciences
90S332375BMB |
Crimp Cap | 75 mm | 20 mL | Modified Top and Bottom |
6800.00 DKK
Pack of 1000 |
Estimated Shipment: 19-01-2026
Log in to see stock. |
||
11873023
|
DWK Life Sciences
90S332375B |
Crimp Cap | 75 mm | 20.0 mL | Modified Top |
7020.00 DKK
Pack of 1000 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||
11853023
|
DWK Life Sciences
90S332346B |
Crimp Cap | 46 mm | 10.0 mL | Modified Top |
5795.00 DKK
Pack of 1000 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||
11863023
|
DWK Life Sciences
90S332375 |
Flat Top Crimp | 75 mm | 20.0 mL | Flat Top |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Finish: 20mm A/S
Order Info
100 vials/pack, 10 packs/case.
Specifications
Clear | |
Silica Glass | |
1000 Cs. | |
20 mm |
23 mm | |
1000 | |
OEM |