missing translation for 'onlineSavingsMsg'
Learn More
Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v) Product Code.: 15484995

Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v)

Product Code. 15484995
4 L, 4 L
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 15484995

Brand: Thermo Scientific Alfa Aesar 045844.K7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

CAS: 64-17-5 | C2H6O | 46.069 g/mol

Denatured ethanol is acting as a solvent to remove the paint and ink stain on curtains, dress and upholstery. It is utilized as fuel in camping and primus stoves. It is likewise employed to clean the windows, manufacturing nail-polish and to test pectin levels in jam and jelly making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -130°C
Color Colorless
Density 0.789 g/mL
Boiling Point 78°C
Quantity 4 L
Flash Point 8°C (46°F)
Physical Form Liquid
Refractive Index 1.361
UN Number UN1170
Beilstein 1718733
Merck Index 14,3760
Solubility Information Miscible with water and most organic solvents.
Formula Weight 46.08
Percent Purity 91.6%
Concentration or Composition (by Analyte or Components) 3.7% methanol, 1% ethyl acetate, 1% toluene (v/v)
Chemical Name or Material Ethanol, Denatured
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ03K033 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog Number045844.K7,045844.M1,045844.K2
Lot NumberT03K023 Certificate TypeCertificate of Analysis Date24/07/2024 Catalog Number045844.K7,045844.M1,045844.K2
Lot NumberS14K039 Certificate TypeCertificate of Analysis Date01/07/2024 Catalog Number045844.K7,045844.M1,045844.K2
Lot NumberR03K040 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number045844.K7,045844.M1,045844.K2
Lot NumberY28J020 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number045844.K7,045844.M1,045844.K2
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H371-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.