Learn More
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | |||||
10652662
|
Lenz Laborglasintrumente
03022473 |
2 L | NS 24/29 |
4570.00 DKK
Pack of 10 |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||||
10758081
|
Lenz Laborglasintrumente
03021946 |
200 mL | Socket NS 19/26 |
1695.00 DKK
Pack of 10 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
11933371
|
Lenz Laborglasintrumente
03021943 |
150 mL | NS 19/26 |
2080.00 DKK
Pack of 10 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
11943371
|
Lenz Laborglasintrumente
03022443 |
150 mL | Socket NS 24/29 |
1950.00 DKK
Pack of 10 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
11953371
|
Lenz Laborglasintrumente
03022943 |
150 mL | Socket NS 29/32 |
1455.00 DKK
Pack of 10 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
Description
- Acc. to DIN EN ISO 4797 and in addition to standard
- With scale (approx. volume /ml)
- With inscription label
- Material: DURAN™
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasintrumente Erlenmeyer Flasks With Conical Ground Joint