Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks With Conical Ground Joint
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03021946
 View more versions of this product
Product Code. 10758081
  • 1695.00 DKK / Pack of 10
Only null left
Estimated Shipment: 24-07-2025
Description
  • Acc. to DIN EN ISO 4797 and in addition to standard
  • With scale (approx. volume /ml)
  • With inscription label
  • Material: DURAN™
Specifications
Specifications
200 mL
Duran Glass
Socket NS 19/26
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasintrumente Erlenmeyer Flasks With Conical Ground Joint > Socket NS 19/26