Learn More
Memmert™ Entry Port Left (Individual Location)
Entry port flap closure custom TBC position in left side
Brand: Memmert™ HPP1400ecoF4
This item is not returnable.
View return policy
Description
- Entry port flap closure custom TBC position in left side
- Can be closed by the flap
Specifications
Clear | |
Not in combination with illumination units, moisture tight, can be closed by flap and silicone stopper | |
HPP1400eco | |
23 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Product Content Correction
Product Title
Memmert™ Entry Port Left (Individual Location) > HPP1400eco
Spot an opportunity for improvement?Share a Content Correction