missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Test Solution Potassium Chloride, SI Analytics™

SI Analytics™ Conductivity Test Solution Potassium Chloride allows for the determination of a substances ability to conduct electricity.

273.00 DKK - 2175.00 DKK

Specifications

Odor Odorless
Density 1g/cm3
Physical Form Fluid
Color Colorless
pH 7
Melting Point 0°C
Boiling Point 100°C
Solubility Miscible with water
View More Specs

Products 4
Product Code Brand Quantity Packaging Concentration Price Quantity & Availability  
Product Code Brand Quantity Packaging Concentration Price Quantity & Availability  
11914809
View Documents
SI Analytics
285126293
20 mL 3 x 6 Ampules 0.01/0.1/1 mol/L (1.41/12.9/112mS/cm)
427.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
11994779
View Documents
SI Analytics
285126511
20 mL 3 x 6 Ampules 0.01 mol/L
427.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
12037112
View Documents
SI Analytics
285136907
250 mL Polyethylene Bottle 0.01 mol/L
273.00 DKK
250mL
Please to purchase this item. Need a web account? Register with us today!
10501603
View Documents
SI Analytics
285126540
500 mL Polyethylene Bottle 5mS/cm
2175.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Mixture consisting of water and potassium chloride
  • Store at room temperature
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Product does not present an explosion hazard
  • Do not allow undiluted product or large quantities of it to reach ground water, water course or sewage system
  • Small amounts may be diluted with plenty of water and washed away
Specifications

Specifications

Odorless
Fluid
7
100°C
1g/cm3
Colorless
0°C
Miscible with water
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Conductivity Test Solution Potassium Chloride, SI Analytics™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.