missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Test Solution Potassium Chloride, SI Analytics™
Shop All SI Analytics Products
Click to view available options
Quantity:
20 mL
250 mL
500 mL
Packaging:
3 x 6 Ampules
Polyethylene Bottle
Concentration:
0.01 mol/L
0.01/0.1/1 mol/L (1.41/12.9/112mS/cm)
5mS/cm
Unit Size:
250mL
Each
Description
- Mixture consisting of water and potassium chloride
- Store at room temperature
- The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
- Product does not present an explosion hazard
- Do not allow undiluted product or large quantities of it to reach ground water, water course or sewage system
- Small amounts may be diluted with plenty of water and washed away

Specifications
Specifications
Packaging | 3 x 6 Ampules |
Odor | Odorless |
Density | 1g/cm3 |
Physical Form | Fluid |
Concentration | 0.01 mol/L |
Color | Colorless |
pH | 7 |
Melting Point | 0°C |
Boiling Point | 100°C |
Solubility | Miscible with water |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Conductivity Test Solution Potassium Chloride, SI Analytics™ >
Spot an opportunity for improvement?Share a Content Correction