missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffers for NucleoSpin™ Virus Kits

Supplemental buffer solutions (e.g., wash and lysis buffers) for use with NucleoSpin™ Virus kits for the rapid isolation of viral nucleic acids (RNA and DNA).

333.00 DKK - 1610.00 DKK

Specifications

Color Colorless
For Use With (Equipment) Compatible NucleoSpin Virus kits
Odor Odorless
For Use With (Application) Isolation of viral RNA and DNA
View More Specs

Products 3
Product Code Brand Quantity Product Type Price Quantity & Availability  
Product Code Brand Quantity Product Type Price Quantity & Availability  
15849137
View Documents
Macherey-Nagel™
740925
120 mL Lysis Buffer RAV1
1610.00 DKK
120mL
Please to purchase this item. Need a web account? Register with us today!
15869137
View Documents
Macherey-Nagel™
740928.40
40 mL RAV1 + 1.0 mg Carrier RNA Lysis Buffer RAV1 + Carrier RNA
682.00 DKK
40mL
Please to purchase this item. Need a web account? Register with us today!
15859137
View Documents
Macherey-Nagel™
740926.1
200 mL Wash Buffer RAV3
333.00 DKK
200mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Compatible kits include:

  • NucleoSpin™ RNA Virus
  • NucleoSpin™ RNA Virus F
  • NucleoSpin™ Dx Virus
  • NucleoSpin™ 8/96 Virus
Specifications

Specifications

Colorless
Compatible NucleoSpin Virus kits
Odorless
Isolation of viral RNA and DNA
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : At least 1 year, in original packaging (prepared/working solutions have a shorter shelf life)

Product Identifier
  • Buffer RAV1 (120 mL)

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Supplemental information
  • MIXTURE LIST-Contains : guanidine thiochloride, triton x-100

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Buffers for NucleoSpin™ Virus Kits

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.