Learn More
Ansell BioClean™ Synergy Non-Sterile Nitrile Gloves
Non-Sterile nitrile gloves offer resistance to a range of chemicals, feature easy double-donning, and have low particle count.
3445.00 DKK
Specifications
Accelerator Free | Yes |
---|---|
Color | Green |
Latex Free | No |
Material | Nitrile |
Sterility | Non-sterile |
Alert:
Warning: No glove provides complete protection against all chemicals. Users must test the gloves against the particular chemicals and environment in which they will be used.
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15208228
![]() |
Ansell
BSAN-M |
Medium |
3445.00 DKK
Pack of 1000 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||||
15298218
![]() |
Ansell
BSAN-S |
Small |
3445.00 DKK
Pack of 1000 |
Estimated Shipment: 12-08-2025
Log in to see stock. |
||||||
15228228
![]() |
Ansell
BSAN-XL |
X-Large |
3445.00 DKK
Pack of 1000 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
15218228
![]() |
Ansell
BSAN-L |
Large |
3445.00 DKK
Pack of 1000 |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
15238228
![]() |
Ansell
BSAN-XXL |
2X-Large |
3445.00 DKK
Pack of 1000 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
Description
- Resistant to a range of chemicals
- Powder, sulphur and accelerator free
- Beaded cuff
- Easy double-donning
- Non-particulating EasyTear packaging
- Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications
- Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425
- Processed in an NEBB certified ISO Class 4 environment
- Complies with the requirements of European Glove Standards EN420:2003 +A1:2009, EN374-1:2016, EN374-5:2016 and EN421:2010
- Physical properties comply with European Medical Glove Standard EN455-2:2015
- Suitable for contact with dry, alcoholic and acidic foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006
Specifications
Yes | |
No | |
Non-sterile | |
12 in. |
Green | |
Nitrile | |
100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces) | |
30 cm |
Five (5) years from date of manufacture. Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.