missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOWA™ 707D Chemical-Protection Nitrile Gloves
The 707D chemical-protection gloves combine the best of disposable and chemical-resistant technology. This hybrid results in the ideal solution for jobs requiring contact with chemical hazards, optimum comfort, and tactility.
169.00 DKK - 177.00 DKK
Specifications
Certifications/Compliance | 21 CFR 170-199, ANSI/ISEA 105-2005 |
---|---|
Color | Blue |
Latex Free | Yes |
Material | Nitrile |
Thickness (English) | 9 mil |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17877311
|
SHOWA™
707D-10XL |
X-Large |
177.00 DKK
12 pairs |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
17887311
|
SHOWA™
707D-11XXL |
2X-Large | N/A | |||||||
17867311
|
SHOWA™
707D-06XS |
X-Small | N/A | |||||||
17871191
|
SHOWA™
707D-08M |
Medium |
169.00 DKK
12 pairs |
Estimated Shipment: 08-09-2025
Log in to see stock. |
||||||
17881191
|
SHOWA™
707D-09L |
Large |
177.00 DKK
12 pairs |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
Description
- Form-fitting
- Easy donning and doffing
- Chemical-resistant
- Oil-resistant
- Water-resistant
- Abrasion-resistant
- Hydrocarbon-resistant
- Impermeable
- Biodegradable
- Ergonomic
- Rolled Cuff
- 100% Nitrile
- Chlorinated
- Embossed grip
- Unlined
- Eco Best Technology™ (EBT)
- Industries: Chemical, Food, Janitorial, Laboratory, Mining, Pharmaceutical
Specifications
21 CFR 170-199, ANSI/ISEA 105-2005 | |
Yes | |
9 mil | |
Rolled | |
12 in. |
Blue | |
Nitrile | |
manufacturing, food processing, laboratories, janitorial, paint applications, poultry processing, light duty maintenance and clean up | |
Rough | |
30.5 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SHOWA™ 707D Chemical-Protection Nitrile Gloves