Learn More
Sartorius Arium™ Advance L Reverse Osmosis System
Arium™ Advance L Reverse Osmosis System for the production of pure water.
120375.00 DKK - 148615.00 DKK
Specifications
Feedwater Source | Potable Tap Water |
---|---|
Product Water Type | Reverse Osmosis (Type 3) |
Type | Water System |
Feedwater Connector | 0.25 in. |
Resistivity | 0.05 MΩ*cm |
Product Code | Brand | Flow Rate | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Capacity (Metric) | Price | Quantity & Availability | |||||
16857296
|
Sartorius
H2O-RO-L-100 |
100 L/hr | 100 L |
120375.00 DKK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
16837296
|
Sartorius
H2O-RO-H-100 |
200 L/hr | 100 L |
130845.00 DKK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
16867296
|
Sartorius
H2O-RO-L-200 |
100 L/hr | 200 L |
138225.00 DKK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
16847296
|
Sartorius
H2O-RO-H-200 |
200 L/hr | 200 L |
148615.00 DKK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
Description
- The Arium™ Advance L laboratory water systems are designed for a reverse osmosis water requirement of up to 200 L/hr
- Combined with the integrated 100 or 200 liter storage tank the system provides the ideal volume of pre-treated feed water for your laboratory.
- The system can simultaneous feed several laboratory instruments at once, thanks to the powerful distributor pump.
- Its compact, square shape reduces space requirements to a minimum.
Specifications
Potable Tap Water | |
Water System | |
0.05 MΩ*cm |
Reverse Osmosis (Type 3) | |
0.25 in. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sartorius Arium™ Advance L Reverse Osmosis System