Learn More
Sartorius Arium™ Advance L Reverse Osmosis System
Arium™ Advance L Reverse Osmosis System for the production of pure water.
Brand: Sartorius H2O-RO-H-100
123031.24 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- The Arium™ Advance L laboratory water systems are designed for a reverse osmosis water requirement of up to 200 L/hr
- Combined with the integrated 100 or 200 liter storage tank the system provides the ideal volume of pre-treated feed water for your laboratory.
- The system can simultaneous feed several laboratory instruments at once, thanks to the powerful distributor pump.
- Its compact, square shape reduces space requirements to a minimum.
Specifications
100 L | |
200 L/hr | |
Water System | |
0.05 MΩ*cm |
Potable Tap Water | |
Reverse Osmosis (Type 3) | |
0.25 in. |
Product Content Correction
Product Title
Sartorius Arium™ Advance L Reverse Osmosis System > 200 L/hr; 100 L
Spot an opportunity for improvement?Share a Content Correction