Learn More
Ansell™ AlphaTec™ 2000 Ts PLUS Stitched and Taped
AlphaTec™ 2000 Ts PLUS Stitched and taped is designed to allow water vapor (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals and filter 100% of particulates down to 0.01 microns in size.
4475.00 DKK - 4940.00 DKK
Specifications
Product Type | Controlled Environments ESD Coveralls |
---|---|
Gender | Unisex |
Material | Microporous Non-woven Polyethylene Laminate |
Color | White |
Disposable | Reusable |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
16844820
|
Ansell™
WH20T-00156-02 |
Small | N/A | |||||||
16854820
|
Ansell™
WH20T-00156-03 |
Medium |
4940.00 DKK
Pack of 30 |
Estimated Shipment: 30-10-2025
Log in to see stock. |
||||||
16864820
|
Ansell™
WH20T-00156-04 |
Large | N/A | |||||||
16874820
|
Ansell™
WH20T-00156-05 |
X-Large | N/A | |||||||
16884820
|
Ansell™
WH20T-00156-06 |
2X-Large | N/A | |||||||
16894820
|
Ansell™
WH20T-00156-07 |
3X-Large |
4940.00 DKK
Pack of 30 |
Estimated Shipment: 30-10-2025
Log in to see stock. |
||||||
Description
Designed to allow perspiration to escape from the suit and withstand saturation of liquid chemicals.
- Filters 100% of particulates down to 0.01 microns in size
- Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric to reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static, tested according to EN 1149-5
- Ultra Low Linting, reduced risk of contamination in critical areas
- Silicone free, critical in spray painting applications
- Tunnelled elasticated hood, wrists, and ankles to minimize the risk of linting and cross contamination
Specifications
Controlled Environments ESD Coveralls | |
Microporous Non-woven Polyethylene Laminate | |
Reusable | |
Hooded | |
Stitched, Taped | |
CE CATEGORY III | |
CE CATEGORY III (Type 4-B, Type 5-B, Type 6-B, EN 1073-2, EN 14126, DIN 32781, EN 1149-5) |
Unisex | |
White | |
Elastic | |
Zipper | |
Non-sterile | |
Hood | |
None |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.