Learn More
Ansell AlphaTec™ 2000 STANDARD Lab Coat Bound
AlphaTec™ 2000 STANDARD Lab coat is designed to allow water vapour (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals and filter 100% of particulates down to 0.01 microns in size.
3605.00 DKK
Specifications
Product Type | Controlled Environments Lab Coat |
---|---|
Gender | Unisex |
Material | Microporous Non-woven Polyethylene Laminate |
Color | White |
Closure Type | Zipper |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
16824790
|
Ansell
WH20B-00231-02 |
Small |
3605.00 DKK
Pack of 60 |
Estimated Shipment: 19-11-2025
Log in to see stock. |
||||||
16834790
|
Ansell
WH20B-00231-03 |
Medium |
3605.00 DKK
Pack of 60 |
Estimated Shipment: 19-11-2025
Log in to see stock. |
||||||
16844790
|
Ansell
WH20B-00231-04 |
Large |
3605.00 DKK
Pack of 60 |
Estimated Shipment: 15-08-2025
Log in to see stock. |
||||||
16854790
|
Ansell
WH20B-00231-05 |
X-Large |
3605.00 DKK
Pack of 60 |
Estimated Shipment: 09-12-2025
Log in to see stock. |
||||||
16864790
|
Ansell
WH20B-00231-06 |
2X-Large |
3605.00 DKK
Pack of 60 |
Estimated Shipment: 19-11-2025
Log in to see stock. |
||||||
Description
Designed to allow perspiration to escape from the suit and withstand saturation of liquid chemicals.
- Filter 100% of particulates down to 0.01 microns in size
- Proven protective barrier to low concentration liquid chemicals, liquids, and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
- Optimized fit, improving wearer comfort and safety
- Low linting
Specifications
Controlled Environments Lab Coat | |
Microporous Non-woven Polyethylene Laminate | |
Zipper | |
Knee-length | |
CE CATEGORY III (Type PB (6)-B, EN 14126) |
Unisex | |
White | |
Reusable | |
2000 | |
Not Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell AlphaTec™ 2000 STANDARD Lab Coat Bound