Learn More
SHOWA™ 477 Series Nitrile Coated Cold Resistant Gloves
Designed to keep hands warm and dry in the most extreme environments, the 477 cold-resistant glove combines thermal insulation with tactical grip.
7370.00 DKK - 7820.00 DKK
Specifications
Certifications/Compliance | CE Cat II, EN 388:2016, EN 511:2006 |
---|---|
Construction | Seamless Knit |
Coating Material | Nitrile |
Color | Blue, Black |
Material | Nylon, Polyester |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17717617
|
SHOWA™
477-07M |
Medium |
7665.00 DKK
5 pairs |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
17707617
|
SHOWA™
477-08L |
Large |
7820.00 DKK
5 pairs |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
17727617
|
SHOWA™
477-09XL |
X-Large |
7370.00 DKK
5 pairs |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
17737617
|
SHOWA™
477-10XXL |
2X-Large |
7370.00 DKK
5 pairs |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
Description
Benifits
- Seamless knit
- Cold Resistant
- Water-resistant
- Flexible
- Abrasion-resistant
- Extra Grip
- Thermal insulation
- Form-fitting
- Impermeable
Features
- Foam grip
- Elasticated wrist cuff
- Ergonomic
- Foam nitrile palm dip
- Insulated
- Fully-coated nitrile
Trades & Applications
- Concreting
- Guttering and drain pipe
- Insulation
- Washing and cleaning
- Planting & repotting
- Bottling
- Water/Sewage treatment
- Plumbing
- Drainage, piping
- Shipping/Receiving
Specifications
CE Cat II, EN 388:2016, EN 511:2006 | |
Nitrile | |
Nylon, Polyester | |
Cold Resistant, Water Resistant, Abrasion Resistant |
Seamless Knit | |
Blue, Black | |
Acrylic | |
477 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
SHOWA™ 477 Series Nitrile Coated Cold Resistant Gloves