Learn More
SHOWA™ 477 Series Nitrile Coated Cold Resistant Gloves
Designed to keep hands warm and dry in the most extreme environments, the 477 cold-resistant glove combines thermal insulation with tactical grip.
Brand: SHOWA™ 477-09XL
This item is not returnable.
View return policy
Description
Benifits
- Seamless knit
- Cold Resistant
- Water-resistant
- Flexible
- Abrasion-resistant
- Extra Grip
- Thermal insulation
- Form-fitting
- Impermeable
Features
- Foam grip
- Elasticated wrist cuff
- Ergonomic
- Foam nitrile palm dip
- Insulated
- Fully-coated nitrile
Trades & Applications
- Concreting
- Guttering and drain pipe
- Insulation
- Washing and cleaning
- Planting & repotting
- Bottling
- Water/Sewage treatment
- Plumbing
- Drainage, piping
- Shipping/Receiving
Specifications
CE Cat II, EN 388:2016, EN 511:2006 | |
Nitrile | |
Nylon, Polyester | |
Acrylic | |
477 |
Seamless Knit | |
Blue, Black | |
X-Large | |
Cold Resistant, Water Resistant, Abrasion Resistant |
Product Content Correction
Product Title
SHOWA™ 477 Series Nitrile Coated Cold Resistant Gloves > X-Large
Spot an opportunity for improvement?Share a Content Correction