Learn More
4-Dimethylaminobenzaldehyde, ACS Product Code.: 11334207
4-Dimethylaminobenzaldehyde, ACS
Product Code. p-7050390
25 g, 25g
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. 11334207
Brand: Thermo Scientific Alfa Aesar 036684.14
Request Bulk or Custom Format
CAS: 100-10-7 | C9H11NO | 149.19 g/mol
4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonalShow MoreShow Less
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1
Specifications
Melting Point 73°C to 75°C
Color White to Yellow
Boiling Point 176°C to 177°C (17 mmHg)
Flash Point 164°C (327°F)
Odor Strong
Quantity 25 g
Beilstein 606802
Sensitivity Air and light sensitive
Merck Index 14,3230
Solubility Information Soluble in alcohol,ether,chloroform,acetic acid; Insoluble in water.
Formula Weight 149.19
Grade ACS
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzaldehyde
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberS09L005 Certificate TypeCertificate of Analysis Date10/06/2025 Product Code036684.14,036684.36,036684.22
Lot NumberU05K055 Certificate TypeCertificate of Analysis Date16/08/2024 Product Code036684.14,036684.36,036684.22
Lot NumberU03G040 Certificate TypeCertificate of Analysis Date02/02/2024 Product Code036684.14,036684.36,036684.22
Lot NumberZ19I026 Certificate TypeCertificate of Analysis Date11/01/2023 Product Code036684.14,036684.36,036684.22
Lot NumberW22I026 Certificate TypeCertificate of Analysis Date13/10/2022 Product Code036684.14,036684.36,036684.22
RUO – Research Use Only
Product Content Correction
Product Title
4-Dimethylaminobenzaldehyde, ACS >