Få mere at vide
2-(4-Methylphenyl)propan-2-ol, TRC
Artikelnummer. 30478912
Skift visning
Mængde:
1 g
2.5 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30478912 5 g
30473206 1 g
30469791 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30478912 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3277155G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1197-01-9
Molekylær formel C10H14O
Molekylvægt (g/mol) 150.22
Synonym 1-Methyl-4-(1-hydroxy-1-methylethyl)benzene, 1-Methyl-4-(alpha-hydroxyisopropyl)benzene, 2-(4-Methylphenyl)-2-propanol, 2-(p-Methylphenyl)-2-propanol, 2-(p-Tolyl)-2-propanol, 4-Methylcumyl alcohol, 8-Hydroxy-p-cymene, Dimethyl-p-tolyl carbinol, p-Cymene-8-ol, alpha,alpha,4-Trimethylbenzyl alcohol
IUPAC navn 2-(4-methylphenyl)propan-2-ol
SMIL Cc1ccc(cc1)C(C)(C)O
Tekniske data
Kemisk navn eller materiale 2-(4-Methylphenyl)propan-2-ol
Mængde 5 g
InChI formel InChI=1S/C10H14O/c1-8-4-6-9(7-5-8)10(2,3)11/h4-7,11H,1-3H3
Formel vægt 150.1045