Thermo Scientific GENESYS Offer
Exclusive to New Lab Start-Up Customers
One Time Offer: Save 25% on your Genesys Spectrophotometer
Valid until 31st December 2024.
Thermo Scientific™ GENESYS™ 50 UV-Vis Spectrophotometer
Catalog No. 16604932
The GENESYS 50 is a dual beam UV-Vis spectrophotometer in a single-cell configuration that meets all of the specifications for critical quality control applications and precise chemical measurements.
Perform routine measurements for quality control or teaching in the chemistry classroom with the rugged Thermo Scientific™ GENESYS™ 50 UV-Visible Spectrophotometer. Offered in a single cell configuration, these instruments are designed with a sloping surface to shed spills and a washable sampling compartment for easy clean up. A 7-inch touchscreen interface, local control software (no PC needed) and optional Wi-Fi networking make the GENESYS 50 UV-Vis Spectrophotometer the ideal choice for daily sampling purposes.
Shop Now
Thermo Scientific™ GENESYS™ 150 UV-Vis Spectrophotometer
Catalog No. 16614932
The GENESYS 150 UV-Vis Spectrophotometer provides automation for high-throughput options and room-light resistance, allowing lid-open operation for improved speed and convenience.
A full range of accessories, including automated cell changers and cell holders, sipper, and fiber optic probes are designed to simplify sampling and accommodate high throughput measurements and temperature control. Features data resolution from 0.2 nm to 5 nm.
Shop NowFill out the form below to receive a promotional code and benefit from this offer.