accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 86953-79-9

CH3H3CCH3OON

CAS RN 86953-79-9

IUPAC Name: tert-butyl pyrrolidine-1-carboxylate
Synonyms: tert-butyl pyrrolidine-1-carboxylate
Molecular Weight (g/mol): 171.24
Molecular Formula: C9H17NO2
InChi Key: LPQZERIRKRYGGM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCCC1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.