accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 51013-67-3

CAS RN 51013-67-3

IUPAC Name: 4-[(4-aminophenyl)methyl]morpholin-4-ium
Synonyms: 4-[(4-aminophenyl)methyl]morpholin-4-ium
Molecular Weight (g/mol): 193.27
Molecular Formula: C11H17N2O
InChi Key: WNYFVEFUHMDIRQ-UHFFFAOYSA-O
SMILES: NC1=CC=C(C[NH+]2CCOCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.