CAS RN 40321-99-1
CAS RN 40321-99-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Mono(2-ethyl-5-hydroxyhexyl) Phthalate (Mixture of Diastereomers) (>90%), TRC
CAS: 40321-99-1 Molekylær formel: C16 H22 O5 Molekylvægt (g/mol): 294.34 Synonym: 1,2-Benzenedicarboxylic acid, mono(2-ethyl-5-hydroxyhexyl) ester (9CI),Mono-2-ethyl-5-hydroxyhexyl phthalate IUPAC navn: 2-(2-ethyl-5-hydroxyhexoxy)carbonylbenzoic acid SMIL: CCC(CCC(C)O)COC(=O)c1ccccc1C(=O)O
2
Phthalic acid, mono-2-ethyl-5-hydroxyhexyl ester 100 μg/mL in Methyl-tert-butyl ether, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality