CAS RN 24522-30-3
IUPAC Navn:
2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Synonymer:
2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Molekylvægt (g/mol):
228.17
Molekylformel:
C10H7F3N2O
InChi Key:
JBNCFFDGYDZEEN-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(NC(=O)CC#N)C=C1
1
–
2
af
2
Resultater
1
2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide, TRC
CAS: 24522-30-3 Molekylær formel: C10 H7 F3 N2 O Molekylvægt (g/mol): 228.17 Synonym: 2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide,Leflunomide Imp. H (EP) IUPAC navn: 2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide SMIL: FC(F)(F)c1ccc(NC(=O)CC#N)cc1
2
2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.