CAS RN 22255-22-7
CAS RN 22255-22-7
IUPAC Name:
1,3-dimethoxy-5-[2-(4-methoxyphenyl)ethenyl]benzene
Synonyms:
1,3-dimethoxy-5-[2-(4-methoxyphenyl)ethenyl]benzene
Molecular Weight (g/mol):
270.33
Molecular Formula:
C17H18O3
InChi Key:
GDHNBPHYVRHYCC-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=CC2=CC(OC)=CC(OC)=C2)C=C1
1
–
1
of
1
results
1
Medchem Express trans-Trimethoxyresveratrol, MedChemExpress
MedChemExpress Trans-Trimethoxyresveratrol is a derivative of Resveratrol (RSV),and it may be a more potent anti-inflammatory, antiangiogenic and vascular-disrupting agent when compared with resveratrol.
