CAS RN 141625-93-6
CAS RN 141625-93-6
IUPAC Name:
hydrogen N-(2-butyl-3-{4-[3-(dibutylamino)propoxy]benzoyl}-1-benzofuran-5-yl)methanesulfonamide chloride
Synonyms:
hydrogen multaq chloride
Molecular Weight (g/mol):
593.22
Molecular Formula:
C31H45ClN2O5S
InChi Key:
DWKVCQXJYURSIQ-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CCCCN(CCCC)CCCOC1=CC=C(C=C1)C(=O)C1=C(CCCC)OC2=CC=C(NS(C)(=O)=O)C=C12
1
–
1
of
1
results
1
Medchem Express Dronedarone Hydrochloride, MedChemExpress
MedChemExpress Dronedarone Hydrochloride is a non-iodinated amiodarone derivative that inhibits Na+, K+ and Ca2+ currents.
