CAS RN 127-48-0
CAS RN 127-48-0
IUPAC Navn:
trimethyl-1,3-oxazolidine-2,4-dione
Synonymer:
trimethadione pitmal petimalin neo-absentol tricione trilidona troxidone trimethadion trimethin trimethadionum
Molekylvægt (g/mol):
143.14
Molekylformel:
C6H9NO3
InChi Key:
IRYJRGCIQBGHIV-UHFFFAOYSA-N
SMILES:
CN1C(=O)OC(C)(C)C1=O
1
–
2
af
2
Resultater
1
Trimethadione, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Trimethadione, MedChemExpress
MedChemExpress Trimethadione (3,5,5,-Trimethyloxazolidine-2,4-dione) is an oxazolidinedione anticonvulsant agent widely used against absences seizures. Trimethadione also is a T-type calcium channel blocker which has antihyperalgesic effects.
