CAS RN 77385-90-1
IUPAC Navn:
ethyl 2-(dibenzylamino)acetate
Synonymer:
ethyl 2-(dibenzylamino)acetate
Molekylvægt (g/mol):
283.37
Molekylformel:
C18H21NO2
InChi Key:
AFMDCFOWHWNQBP-UHFFFAOYSA-N
SMILES:
CCOC(=O)CN(CC1=CC=CC=C1)CC1=CC=CC=C1
1
–
3
af
3
Resultater
1
N,N-Dibenzylglycine Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N,N-Dibenzylglycine Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N,N-Dibenzylglycine Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.