CAS RN 764661-24-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
6
af
6
Resultater
1
4-(Methylnitrosamino)-1-(3-pyridyl-d4)-1-butanone-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(Methylnitrosamino)-1-(3-pyridyl-d4)-1-butanone-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
4-(Methylnitrosamino)-1-(3-pyridyl-d4)-1-butanone-d4, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
6
4-(N-Methyl-N-nitrosoamino)-1-(3-pyridyl-d4)-1-butanone, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.