CAS RN 72648-46-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Androsta-1,4-diene-3,6,17-trione, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
6-Oxo Boldione, TRC
CAS: 72648-46-5 Molekylær formel: C19 H22 O3 Molekylvægt (g/mol): 298.38 Synonym: Androsta-1,4-diene-3,6,17-trione (9CI),FCE 24204,6-Oxo Boldione,Androsta-1,4-diene-3,6,17-trione IUPAC navn: (8R,9S,10R,13S,14S)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,6,17-trione SMIL: C[C@]12CC[C@H]3[C@@H](CC(=O)C4=CC(=O)C=C[C@]34C)[C@@H]1CCC2=O