CAS RN 67469-75-4
IUPAC Navn:
1-[2-(diphenylmethoxy)ethyl]-4-(3-phenylprop-2-en-1-yl)piperazine dihydrochloride
Synonymer:
1-[2-(diphenylmethoxy)ethyl]-4-(3-phenylprop-2-en-1-yl)piperazine dihydrochloride
Molekylvægt (g/mol):
485.49
Molekylformel:
C28H34Cl2N2O
InChi Key:
HJDXBLLTRMCYNC-UHFFFAOYSA-N
SMILES:
Cl.Cl.C(CN1CCN(CC=CC2=CC=CC=C2)CC1)OC(C1=CC=CC=C1)C1=CC=CC=C1
1
–
4
af
4
Resultater
1
GBR 12783 Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tocris Bioscience™ GBR 12783 dihydrochloride
CAS: 67469-75-4 Molekylær formel: C28H34Cl2N2O
3
GBR 12783 Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
GBR 12783 Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.