CAS RN 6508-43-6
IUPAC Navn:
(9S)-9-(2-hydroxy-4,4-dimethyl-6-oxocyclohex-1-en-1-yl)-3,3-dimethyl-2,3,4,9-tetrahydro-1H-xanthen-1-one
Synonymer:
(9S)-9-(2-hydroxy-4,4-dimethyl-6-oxocyclohex-1-en-1-yl)-3,3-dimethyl-4,9-dihydro-2H-xanthen-1-one
Molekylvægt (g/mol):
366.46
Molekylformel:
C23H26O4
InChi Key:
CCJIUBMPDVWYLU-IBGZPJMESA-N
SMILES:
CC1(C)CC(O)=C([C@@H]2C3=CC=CC=C3OC3=C2C(=O)CC(C)(C)C3)C(=O)C1
1
–
2
af
2
Resultater
1
Tocris Bioscience™ L-152,804
CAS: 6508-43-6 Molekylær formel: C23H26O4
2
L 152804, MedChemExpress
MedChemExpress L 152804 is an orally active and selective neuropeptide Y Y5 receptor (NPY5-R) antagonist, with a Ki of 26 nM for hY5. L 152804 causes weight loss in diet-induced obese mice by modulating food intake and energy expenditure.