CAS RN 61281-37-6
IUPAC Navn:
3,4,5,19-tetramethoxy-9,10-dimethyl-15,17-dioxatetracyclo[10.7.0.0²,⁷.0¹⁴,¹⁸]nonadeca-1(19),2(7),3,5,12,14(18)-hexaene
Synonymer:
3,4,5,19-tetramethoxy-9,10-dimethyl-15,17-dioxatetracyclo[10.7.0.0²,⁷.0¹⁴,¹⁸]nonadeca-1(19),2(7),3,5,12,14(18)-hexaene
Molekylvægt (g/mol):
400.47
Molekylformel:
C23H28O6
InChi Key:
RTZKSTLPRTWFEV-UHFFFAOYNA-N
SMILES:
COC1=CC2=C(C(OC)=C1OC)C1=C(OC)C3=C(OCO3)C=C1CC(C)C(C)C2
1
–
2
af
2
Resultater
2
Schisandrin B, MedChemExpress
MedChemExpress Schisandrin B (γ-Schisandrin) is a dibenzocyclooctadiene derivative isolated from Fructus Schisandrae, has been shown to produce antioxidant effect on rodent liver and heart.