Få mere at vide
Schisandrin B, MedChemExpress
Artikelnummer. 16478858
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16478858 10 mg
16444747 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16478858 Leverandør Medchem Express Leverandørnr. HYN0089
MedChemExpress Schisandrin B (γ-Schisandrin) is a dibenzocyclooctadiene derivative isolated from Fructus Schisandrae, has been shown to produce antioxidant effect on rodent liver and heart.
Schisandrin B (γ-Schisandrin) is a dibenzocyclooctadiene derivative isolated from Fructus Schisandrae, has been shown to produce antioxidant effect on rodent liver and heart.
Tekniske data
Kemisk navn eller materiale Schisandrin B
CAS 61281-37-6
Farve White
Renhedsgrad noter Research
Molekylær formel C23H28O6
Mængde 10 mg
Synonym γ-Schisandrin Wuweizisu B
Opløselighedsinformation DMSO : 100 mg/mL (249.71 mM; Need ultrasonic)
SMIL CC(C1)C(C)CC2=CC(OC)=C(OC)C(OC)=C2C3=C1C=C4OCOC4=C3OC
Molekylvægt (g/mol) 400.46
Formel vægt 400.46
Procent renhed 98.98%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death