CAS RN 600-05-5
IUPAC Navn:
2,3-dibromopropanoic acid
Synonymer:
propanoic acid, 2,3-dibromo-
Molekylvægt (g/mol):
231.87
Molekylformel:
C3H4Br2O2
InChi Key:
ZMYAKSMZTVWUJB-UHFFFAOYNA-N
SMILES:
OC(=O)C(Br)CBr
1
–
6
af
6
Resultater
1
2,3-Dibromopropanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,3-Dibromopropanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,3-Dibromopropanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Molybdænopløsning til ICP , 10 g/L, HNO 3 /HF Matrix, Fisher Chemical™
CAS: 600-05-5 Molekylær formel: C3H4Br2O2 Molekylvægt (g/mol): 231.87 MDL nummer: MFCD00004212 InChI nøgle: ZMYAKSMZTVWUJB-UHFFFAOYNA-N PubChem CID: 11746 IUPAC navn: 2,3-dibrompropansyre SMIL: OC(=O)C(Br)CBr