CAS RN 59960-32-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Dihydro Ketoprofen (Mixture of Diastereomers), TRC
CAS: 59960-32-6 Molekylær formel: C16 H16 O3 Molekylvægt (g/mol): 256.3 Synonym: 2-[3-(alpha-Hydroxybenzyl)phenyl]propanoic Acid IUPAC navn: 2-[3-[hydroxy(phenyl)methyl]phenyl]propanoic acid SMIL: CC(C(=O)O)c1cccc(c1)C(O)c2ccccc2
2
2-[3-(alpha-Hydroxybenzyl)phenyl]propanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.