CAS RN 58577-87-0
IUPAC Navn:
(2R)-2-amino-3-(benzyloxy)propan-1-ol
Synonymer:
(2R)-2-amino-3-(benzyloxy)propan-1-ol
Molekylvægt (g/mol):
181.24
Molekylformel:
C10H15NO2
InChi Key:
ZJUOMDNENVWMPL-SNVBAGLBSA-N
SMILES:
N[C@H](CO)COCC1=CC=CC=C1
1
–
4
af
4
Resultater
1
(R)-(+)-2-Amino-3-benzyloxy-1-propanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-(+)-2-Amino-3-benzyloxy-1-propanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(R)-(+)-2-Amino-3-benzyloxy-1-propanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.