CAS RN 5312-73-2
IUPAC Navn:
1-(dichloromethoxy)butane
Synonymer:
1-(dichloromethoxy)butane
Molekylvægt (g/mol):
157.03
Molekylformel:
C5H10Cl2O
InChi Key:
GVVHBNABFJZWIC-UHFFFAOYSA-N
SMILES:
CCCCOC(Cl)Cl
1
–
3
af
3
Resultater
1
Dichloromethyl N-Butyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Dichloromethyl N-Butyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Dichloromethyl N-Butyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.