CAS RN 51963-61-2
IUPAC Navn:
disodium trihydrate hydrogen [({[5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate
Synonymer:
disodium trihydrate hydrogen {[5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy(hydroxy)phosphoryl phosphonato}oxyphosphonate
Molekylvægt (g/mol):
605.19
Molekylformel:
C10H20N5Na2O16P3
InChi Key:
MWEQTWJABOLLOS-UHFFFAOYNA-L
SMILES:
O.O.O.[Na+].[Na+].NC1=C2N=CN(C3OC(COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)C(O)C3O)C2=NC=N1
1
–
3
af
3
Resultater
1
Adenosin-5'-trifosfatdinatriumsalthydrat MP Biomedicals
CAS: 51963-61-2 Molekylær formel: C10H20N5Na2O16P3 Molekylvægt (g/mol): 605.19 MDL nummer: MFCD00150754 InChI nøgle: MWEQTWJABOLLOS-MVHBFMHHNA-L PubChem CID: 134129625 SMIL: O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
2
Melford ATP
Et essentielt nukleotidtriphosphat med en række forskellige roller i energimetabolisme, signaltransduktion og nukleinsyremetabolisme.
3
Adenosine 5'-Triphosphate Disodium Salt Trihydrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.